Order Entry
Poland
ContactUsLinkComponent
VWR®, Ethyl linoleate (neat), CRM
VWR®, Ethyl linoleate (neat), CRM
Catalog # 91651.0100
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Ethyl linoleate (neat), CRM
Catalog # 91651.0100
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule/vial
  • Pk:
    100 mg
  • Standards:
    CRM ISO 17034
  • CAS No.:
    544-35-4
  • Description:
    Ethyl linoleate (neat)
  • Storage conditions:
    Freeze (<−10 °C)/Under N2
  • Formula:
    C20H36O2
  • Molecular weight:
    308,50
  • Description:
    Ethyl linoleate

Specifications

About this item

Certifications: Certified reference material.