VWR®, (S)-(-)-1, 2,4-Butanetriol, 1 mg/ml in pyridine, CRM
Catalog # 91622.050
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule/vial
- Pk:5 mL
- Standards:CRM ISO 17034
- CAS No.:42890-76-6
- Description:(S)-(-)-1, 2,4-Butanetriol, 1 mg/ml in pyridine
- Storage conditions:Refrig (0 - 5 °C)
- UN No.:1282
- Description:(S)-(-)-1,2,4-Butanetriol
Specifications
About this item
Certifications: Certified reference material.
Caution: Highly Flammable.