H Series Laminar Chamber with Horizontal Airflow
Supplier: ALPINA POLSKA
Certificates
About this item
The H series chambers are designed to provide a working environment completely free of particles and bacteria. They create sterile conditions for the processed material, they are recommended for working with samples that do not pose a microbiological risk.
- Horizontal, laminar air flow in the range of 0.25 to 0.54 m/s
- Air flow speed control by sensor and silent fans
- HEPA H14 filter and prefilter class EU3 (G3) polyamide acc. PN-EN 779 standard
- Automatic compensation of filter wear for a constant and safe flow rate in the chambers, hourly counter of device and UV lamp operation
- UV germicidal fluorescent lamp fixed permanently, placed in the upper front part of the working area, protected against accidental switching on during operation
- Fluorescent lamps illuminating the workspace with the option of working in two modes, 800 lux and above 1400 lux
- Stainless steel worktop, sides of the workspace with window openings, electrical socket on the side on the top panel
Intuitive control via a touch screen in Polish. Launching individual functions using dedicated and appropriately marked buttons without having to enter any menu makes work much easier. The use of EC fans and energy-saving lighting reduced power consumption by 68% compared to the previous model.
Delivery: Optional valves for vacuum, gas and air, a blend and a dedicated base on lockable wheels.
Specifications
- Catalog No:
- ALPIH180
- ALPIH120
- ALPIH150
- Ext. W×D×H:
- 1850×790×950 mm
- 1250×790×950 mm
- 1550×790×950 mm