PCR Laminar Chamber with Vertical Airflow
The PCR series chamber has 2 additional UV lamps in the compression chamber, increasing the sterility of the air supplied to the working space.
- Vertical, laminar air flow in the range of 0.25 to 0.50 m/s,
- Switching UV lamps on and off in the compression chamber (PCR - on, PCR - off) from the display
- Air flow velocity control by sensor or volt voltage system with +/- 10% correction by the user
- Quiet fans, HEPA H14 filter and polyamide pre-filter class EU3 (G3) acc. PN-EN 779 standard
- Automatic compensation of filter wear to ensure a constant and safe flow rate in the chambers
- An hour counter of the device and UV lamp operation time
- UV germicidal fluorescent lamp fixed permanently, placed in the upper front part of the working area, protected against accidental switching on during operation
Control over the correct operating parameters is facilitated by a system that monitors errors and informs about the need for a service examination or the upcoming filter replacement date.
: Optional valves for vacuum, gas and air, and a dedicated base on lockable wheels.
- Catalog No:
- ALPIPCRS100
- ALPIPCR1600
- ALPIPCR700
- ALPIPCR1300
- ALPIPCR1000
- Exterior dimensions:
- 1030×738×1000 mm
- 1624×633×718 mm
- 719×500×780 mm
- 1319×500×780 mm
- 1019×500×780 mm
Frequently Bought Together