Order Entry
Poland
ContactUsLinkComponent
Ruthenium(III) chloride hydrate, ReagentPlus®
Corrosive
Irritant
Ruthenium(III) chloride hydrate, ReagentPlus®
Catalog # SIAL206229-5G
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Ruthenium(III) chloride hydrate, ReagentPlus®
Catalog # SIAL206229-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :207.43 g/mol (anhy)
Melting Pt:> 300 °C
Density:3.11 g/cm³ (20 °C)
EINECS:233-167-5
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:14898-67-0
 :20759-14-2
Formula:Cl₃Ru·xH₂O
MDL Number:MFCD00149844