Order Entry
Poland
ContactUsLinkComponent
 
 
 
 
Copper(II) chloride for synthesis, Sigma-Aldrich®
  8.18247.9020
 :  Merck
 :  
 
 
 
Copper(II) chloride for synthesis, Sigma-Aldrich®
  8.18247.9020
 :  Merck
 :  

 

  • Pack type:
    Fibreboard box/case with liner
  • Pk:
    20 kg
  • Seq:
    4241

 

 

  • Solubility:
    620 g/l (20 °C)
  • Melting Point:
    498 °C decomposes
  • Molar Mass:
    134.45 g/mol
  • Density:
    3.39 g/cm3 (25 °C)
  • pH:
    3.5 (50 g/l, H2O, 20 °C)
  • Synonyms:
    Copper dichloride
  • Hill Formula:
    Cl2Cu
  • Chemical Formula:
    CuCl2

 

 

 :
 :134.45 g/mol
 :8,III
 :M-Clarity™ quality level = MQ200
 :2802
 :GHS05
 :GHS07
 :GHS09
 :7447-39-4
 :Cl₂Cu
 :MFCD00010972