Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Methyl-4-hydroxybenzoate ≥98.0% NF

Methyl-4-hydroxybenzoate ≥98.0% NF
 
Methyl-4-hydroxybenzoate ≥98.0% NF
Methyl-4-hydroxybenzoate ≥98.0% NF

 

 

Methylparaben, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Exclamation mark.

 :  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  •  :99-76-3
  •  :HOC₆H₄CO₂CH₃
  •  :MFCD00002352
  •  :152.15 g/mol
  •  :Ambient
 

 

  • Assay (C₈H₈O₃):
    98.0 - 102.0 %
  • Color of Solution:
    To Pass Test
  • Acidity:
    To Pass Test
  • Residue On Ignition:
    max. 0.1 %
  • Related Substances:
    To Pass Test
Product Family Options

 

  • Pack type
    Pk
     
     
  • 500 g
       
       
    1 * 500 g
    1 * 500 g - 808 zł
    808 zł
    1 * 500 g
       808 zł

  • Fibre drum
    50 kg
       
       
    1 * 50 kg
    1 * 50 kg - 11 470 zł
    11 470 zł
    1 * 50 kg
       11 470 zł