
Propyl-4-hydroxybenzoate sodium salt ≥94.0% NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:35285-69-9
:
Formula:C₁₀H₁₁NaO₃
MDL Number:MFCD00016477
:202.19 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (Anhydrous Basis):94.0 - 102.0 %
- Completeness of Solution:To Pass Test
- pH of a 1 in 1000 Solution:9.5 - 10.5
- Water:max. 5.0 %
- Chloride:max. 0.035 %
Specification Test Results
Frequently Bought Together