Order Entry
Poland
ContactUsLinkComponent
Copper(II) chloride dihydrate
Irritant
Environment
Copper(II) chloride dihydrate
Catalog # SIAL467847-50G
Supplier:  Merck
CAS Number:  
Irritant
Environment
Copper(II) chloride dihydrate
Catalog # SIAL467847-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :170.48 g/mol
Merck Index:13,02660
Melting Pt:100 °C
Density:2.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
 :GHS07
 :GHS09
CAS Number:10125-13-0
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674