Ethyl octanoate Analytical standard, reference material, Fluka™
Ethyl octanoate, Analytical standard, Glass Bottle, 1 g
HONCR1910-1G
Ethyl octanoate Analytical standard, reference material, Fluka™
HONCR1910-1G
:
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- Description:Ethyl octanoate
- Cat. No.:HONCR1910-1G
- Formula:C₁₀H₂₀O₂
- Description:Ethyl octanoate
- Storage conditions:RT
- Physical form:Liquid
- MDL No.:MFCD00009552
- Flash point:82 °C
- Melting point:-44.2 °C
- Synonym(s):Ethyl caprylate
- Molecular weight:172.26 g/mol
- CAS No.:106-32-1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



