Order Entry
Poland
Orders LinkContactUsLinkComponent
Ethyl hexanoate Analytical standard, reference material, Fluka™

Ethyl hexanoate Analytical standard, reference material, Fluka™

Ethyl hexanoate, Analytical standard, Glass Bottle, 1 g
  HONCR1907-1G
Ethyl hexanoate Analytical standard, reference material, Fluka™
  HONCR1907-1G
 :  
Ethyl hexanoate Analytical standard, reference material, Fluka™

Ethyl hexanoate, Analytical standard, Glass Bottle, 1 g

 :  
 
318 zł
   
1 * 1 g
 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
 

 

  • Pack type:
    Glass Bottle
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • Description:
    Ethyl hexanoate
  • UN No.:
    3272
  • Cat. No.:
    HONCR1907-1G
  • Formula:
    C₈H₁₆O₂
  • Description:
    Ethyl hexanoate
  • Storage conditions:
    RT
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00009511
  • Flash point:
    44.5 °C
  • Melting point:
    -68.3 °C
  • Synonym(s):
    Caproic acid ethyl ester; Ethyl caproate
  • Molecular weight:
    144.21 g/mol
  • CAS No.:
    123-66-0