Captan Analytical standard, reference material, Fluka™
Captan, Analytical standard, Glass Bottle, 100 mg
Catalog # HONCR1851-100MG
Supplier: Honeywell Chemicals
Captan Analytical standard, reference material, Fluka™
Catalog # HONCR1851-100MG
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- Description:Captan
- UN No.:2811
- Cat. No.:HONCR1851-100MG
- Formula:C₉H₈Cl₃NO₂S
- Description:Captan
- Storage conditions:RT
- Physical form:Solid
- MDL No.:MFCD00041811
- Melting point:173 - 175 °C
- Synonym(s):3a,4,7,7a-tetrahydro-2-[(trichloromethyl)thio]-1H-isoindole-1,3(2H)-dione
- Molecular weight:300.6 g/mol
- CAS No.:133-06-2



