Order Entry
Poland
ContactUsLinkComponent
Norflurazon Analytical standard, reference material, Fluka™
Norflurazon Analytical standard, reference material, Fluka™
Catalog # HONCR1821-100MG
CAS Number:  
Norflurazon Analytical standard, reference material, Fluka™
Catalog # HONCR1821-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    27314-13-2
  • Description:
    Norflurazon
  • Storage conditions:
    Ambient
  • MDL No.:
    MFCD00128055
  • Synonym(s):
    4-Chloro-5-(methylamino)-2-(a,a,a-trifluoro-m-tolyl)-3(2H)-pyridazinone
  • Formula:
    C₁₂H₉ClF₃N₃O
  • Description:
    Norflurazon
  • Physical form:
    Crystalline

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery