Order Entry
Poland
Orders LinkContactUsLinkComponent
Nicotinamide Analytical standard, reference material, Fluka™

Nicotinamide Analytical standard, reference material, Fluka™

Nicotinamide, Analytical standard, Glass Bottle, 250 mg
Catalog # HONCR1815-250MG
Supplier:   Honeywell Chemicals
Nicotinamide Analytical standard, reference material, Fluka™
Catalog # HONCR1815-250MG
CAS Number:  
Nicotinamide Analytical standard, reference material, Fluka™

Nicotinamide, Analytical standard, Glass Bottle, 250 mg

CAS Number:  
Price
192 zł
Log in for availability
1 * 250 mg
Product Details & Documents

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
Specifications

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • Description:
    Nicotinamide
  • Cat. No.:
    HONCR1815-250MG
  • Formula:
    C₆H₆N₂O
  • Description:
    Nicotinamide
  • Storage conditions:
    Ambient
  • Physical form:
    Crystalline
  • MDL No.:
    MFCD00006395
  • Synonym(s):
    Pyridine-3-carboxylic acid amide; Niacinamide; Vitamin B3,
  • CAS No.:
    98-92-0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.