Order Entry
Poland
Orders LinkContactUsLinkComponent
Hexaconazol Analytical standard, reference material, Fluka™

Hexaconazol Analytical standard, reference material, Fluka™

Hexaconazol, Analytical standard, Glass Bottle, 100 mg
Catalog # HONCR1796-100MG
Supplier:   Honeywell Chemicals
Hexaconazol Analytical standard, reference material, Fluka™
Catalog # HONCR1796-100MG
CAS Number:  
Hexaconazol Analytical standard, reference material, Fluka™

Hexaconazol, Analytical standard, Glass Bottle, 100 mg

CAS Number:  
Price
483 zł
Log in for availability
1 * 100 mg
Product Details & Documents

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
Specifications

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • Description:
    Hexaconazol
  • Cat. No.:
    HONCR1796-100MG
  • Description:
    Hexaconazol
  • Storage conditions:
    RT
  • Physical form:
    Solid
  • MDL No.:
    MFCD00144441
  • Melting point:
    112 °C
  • Molecular weight:
    314.21 g/mol
  • CAS No.:
    79983-71-4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.