VWR®, Perfluorooctane sulphonic acid, Certified Reference Material (CRM)
numer katalogowy 91152.001
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass vial
- Pk:100 mg
- Standards:CRM ISO 17025/17034
- CAS No.:1763-23-1
- Description:Perfluorooctane sulphonic acid
- UN No.:3261
- Synonym(s):Per- and polyfluoroalkyl substances
- Formula:C₈HF₁₇O₃S
- Molecular weight:500,13 g/mol
- Description:Perfluorooctane sulphonic acid
Specifications
About this item
Per- and polyfluoroalkyl substances (PFAS) are amongst the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances has never been more important.
VWR® is supplying a comprehensive range of standards and CRMs to enable this environmental analysis.
Certifications: Certified Reference Material (CRM) ISO 17025 / ISO 17034