Order Entry
Poland
ContactUsLinkComponent
Gabapentin Related Compounds, Reference Standard
Gabapentin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Gabapentin Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    64744-50-9
  • UN No.:
    2811
  • Synonym(s):
    2-Azaspiro[4.5]decan-3-one
  • Formula:
    C₉H₁₅NO
  • Description:
    Gabapentin Related Compound A

Specifications

Related Information

Frequently Bought Together