Order Entry
Poland
ContactUsLinkComponent
 
Fluticasone Propionate Acid, Pharmaceutical Analytical Impurities
  1A00700.
 :  USP
 :  
Fluticasone Propionate Acid, Pharmaceutical Analytical Impurities
  1A00700.
 :  USP
 :  

 

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    65429-42-7
  • Description:
    Fluticasone Propionate Acid
  • Synonym(s):
    (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-(propionyloxy)-6,7,8,9,10,11,12,13,14,15, 16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carboxylic acid
  • Description:
    Fluticasone Propionate Acid

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.