Order Entry
Poland
ContactUsLinkComponent
Temazepam Related Compounds, Reference Standard
Temazepam Related Compounds, Reference Standard
Supplier:  USP
Certificates
Temazepam Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    93329-92-1
  • Synonym(s):
    7-Chloro-1,4-dimethyl-5-phenyl-4,5-dihydro-1H-1,4-benzodiazepine-2,3-dione
  • Description:
    Temazepam Related Compound G

Specifications

Frequently Bought Together