Order Entry
Poland
ContactUsLinkComponent
 
Rivaroxaban Desthiophene Analog, Pharmaceutical Analytical Impurities
  1A01510.
 :  USP
 :  
Rivaroxaban Desthiophene Analog, Pharmaceutical Analytical Impurities
  1A01510.
 :  USP
 :  

 

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    1411775-06-8
  • Description:
    Rivaroxaban Desthiophene Analog
  • Synonym(s):
    (R)-4-(4-(5-(aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one
  • Description:
    Rivaroxaban Desthiophene Analog

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.