Pyrazinediethanol, Pharmaceutical Analytical Impurities
Supplier: USP
Certificates
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.
Specifications
- CAS No.:4744-51-8
- Synonym(s):2,2'-(pyrazine-2,5-diyl)bis(ethan-1-ol)
- Description:Pyrazinediethanol
Specifications
Frequently Bought Together