Order Entry
Poland
ContactUsLinkComponent
Oxybutynin Related Compounds, Reference Standard
Oxybutynin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Oxybutynin Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    1508-65-2
  • Description:
    Oxybutynin Chloride
  • Synonym(s):
    Phenylcyclohexylglycolic Acid

Specifications

Frequently Bought Together