N-Acetyl Desloratadine, Pharmaceutical Analytical Impurities
Catalog # 1A01670.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:117796-52-8
- Description:N-Acetyl Desloratadine
- Synonym(s):1-[4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinyl]ethanone
- Description:N-Acetyl Desloratadine
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.