Order Entry
Poland
ContactUsLinkComponent
Mafenide Related Compounds, Reference Standard
Mafenide Related Compounds, Reference Standard
Supplier:  USP
Certificates
Mafenide Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    3240-35-5
  • Synonym(s):
    4-Formylbenzenesulfonamide
  • Description:
    Mafenide Related Compound A

Specifications

Frequently Bought Together