Ethylpyrazinediethanol, Pharmaceutical Analytical Impurities
1A00360.
: USP
:
- Pack type:Vial
- Pk:10 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:86917-74-0
- Description:Ethylpyrazinediethanol
- Synonym(s):2,2'-(3-ethylpyrazine-2,5-diyl)bis(ethan-1-ol)
- Description:Ethylpyrazinediethanol
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.