Desmethoxy Omeprazole, Pharmaceutical Analytical Impurities
numer katalogowy 1A00060.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:110374-16-8
- Description:Desmethoxy Omeprazole
- Synonym(s):2-(((3,5-dimethylpyridin-2-yl)methyl)sulFood Ingredientsnyl)-5-methoxy-1H-benzo[d]imidazole
- Description:Desmethoxy Omeprazole
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.