Order Entry
Poland
ContactUsLinkComponent
Desfluorociprofloxacin Hydrochloride, Pharmaceutical Analytical Impurities
Desfluorociprofloxacin Hydrochloride, Pharmaceutical Analytical Impurities
Catalog # 1A01740.
Supplier:  USP
CAS Number:  
Desfluorociprofloxacin Hydrochloride, Pharmaceutical Analytical Impurities
Catalog # 1A01740.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    2727369-47-1
  • Description:
    Desfluorociprofloxacin Hydrochloride
  • Synonym(s):
    1-cyclopropyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, hydrochloride
  • Description:
    Desfluorociprofloxacin Hydrochloride

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.