Budesonide 9(11)-ene, Pharmaceutical Analytical Impurities
Catalog # 1A00680.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:313474-58-7
- Description:Budesonide 9(11)-ene
- Synonym(s):(6aS,8aS,8bS,11aR,12aS,12bS)-8b-(2-hydroxyacetyl)-6a,8a-dimethyl-10-propyl-1,2,6a,8,8a,8b,11a,12,12a,12b-decahydro-4H-naphtho[2',1':4,5]indeno[1,2-d][1,3]dioxol-4-one
- Description:Budesonide 9(11)-ene
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.