AseptiQuik® S Connectors
Supplier: CPC COLDER
Certificates
About this item
Connectors provide quick and easy sterile connections for small-flow applications, even in non-sterile environments.
- Intuitive three step connection process reduces risk of operator error
- Repeatable and reliable performance
- Audible confirmation of assembly
- Applicable for gamma radiation and autoclave sterilisation
- Temperature range: 4 to 40 °C
The 'FLIP-CLICK-PULL' design of AseptiQuik S enables users to quickly transfer small volumes of media easily with less risk of operator error. The connector's genderless and robust design provides reliable performance without the need for clamps, fixtures or tube welders.
Specifications
- Catalog No:
- CPCCAQS17004HT
- CPCCAQS17002HT
- Max. operating pressure:
- Vacuum...4,1 bar
- Vacuum...4,1 bar