Powered air respirator filter cover for Versaflo™ TR-6580E, Versaflo™
Catalog # MMMATR-6500FC
Supplier: 3M
Specifications
- Description:Respirator filter cover
- Environmentally Preferable:
- Pk:1
Specifications
About this item
The Versaflo™ Filter Cover TR-6500FC is a filter cover that helps to protect Versaflo™ TR-6580E filters from physical damage.
- Protects 3M™ Versaflo™ TR-6580E filter from physical damage
- Can be cleaned and reused
This filter cover safeguards the filter and enables users to wear Versaflo™ Powered Air Turbo through decontamination showers. It can hold a Versaflo™ Spark Arrestor TR-662, or Versaflo™ Prefilter TR-6600 in place for additional protection from larger particles or sparks. The filter cover can be cleaned for reuse.