Order Entry
Poland
ContactUsLinkComponent
Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
undefined
Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
  • Body material:
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • Differential pressure:
  • 6,9 bar (100 psi)
  • 4,1 bar (60 psi)
  • 3,4 bar (50 psi)
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Gear material:
  • PEEK
  • PEEK
  • PEEK
  • Inlet connection:
  • 0,375 NPT(F)
  • 0,375 NPT(F)
  • 0,75 NPT(F)
  • Internal bypass:
  • No
  • No
  • No
  • Max. temperature:
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • Max. viscosity:
  • 1500 cP
  • 1500 cP
  • 2500 cP
  • Min. temperature:
  • –29 °C (–20 °F)
  • –29 °C (–20 °F)
  • –46 °C (–50 °F)
  • Min. viscosity:
  • 0,2 cP
  • 0,2 cP
  • 0,2 cP
  • Outlet connection:
  • 0,375 NPT(F)
  • 0,375 NPT(F)
  • 0,75 NPT(F)
  • Reversible:
  • -
  • -
  • +
  • Seal material:
  • TEV
  • TEV
  • PTFE
  • Suction lift, wet:
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together