Order Entry
Poland
ContactUsLinkComponent
Sodium hydroxide 1 N in aqueous solution
Corrosive
Sodium hydroxide 1 N in aqueous solution
Catalog # HEWL5062-8576
Supplier:  Agilent
CAS Number:  
Corrosive
Sodium hydroxide 1 N in aqueous solution
Catalog # HEWL5062-8576
Supplier:  Agilent
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548