Order Entry
Poland
ContactUsLinkComponent
PTFE tubing, BOLA
Catalog # BOHLS1810-41
Supplier:  Bohlender
undefined
PTFE tubing, BOLA
Catalog # BOHLS1810-41
Supplier:  Bohlender
Specifications
  • Tubing ext.Ø:
    6,35 mm
  • Tubing int.Ø:
    3,96 mm
  • Pk:
    1
Specifications
About this item
Translucent to milky-white appearance.
  • Almost universal chemical resistance
  • Free from extractable agents
  • Physiologically safe
  • Flame retardant according to UL94V0
Perfect tubing for aggressive and pure liquids or gases.