Order Entry
Poland
ContactUsLinkComponent
Half mask with PressToCheck™ P3 R D filters, Force®8, JSP LTD
Half mask with PressToCheck™ P3 R D filters, Force®8, JSP LTD
Catalog # JSPABHT0A3-0L5-N00
Supplier:  JSP
undefined
Half mask with PressToCheck™ P3 R D filters, Force®8, JSP LTD
Catalog # JSPABHT0A3-0L5-N00
Supplier:  JSP
Specifications
  • Description:
    Force®8 Half mask with PressToCheck™ P3 R D filters
  • Pk:
    1
Specifications
About this item
The Force®8 twin cartridge half mask with Typhoon™ valve offers superior low breathing resistance and a 4-point suspension harness with quick-release buckles, fitted with P3 PressToCheck™ filters for daily face fit reassurance.
  • Durable thermoplastic rubber mask for superior fit to most face shapes
  • Fully adjustable 4-point cradle suspension harness
  • Low resistance exhalation valve
  • Fitted with PressToCheck™ P3 R D filters
  • CR2 reflectivity
The Force®8 twin cartridge half mask with Typhoon™ valve is made with a durable thermoplastic rubber, offering a superior fit to most face shapes. The mask is also fitted with PressToCheck™ P3 R D filters for daily face fit reassurance.
CR2 reflective strips create increased visibility in low-light environments for added safety.
Certifications: Conforms to EN 140 & EN 143:200.