T-connection tubes for waste pipes, b.safe
Catalog # BOHLR1190-01
Dostawca: Bohlender
Dane techniczne
- Description:T-connection tubes for waste pipes, PE, with 2× clamp connection Ø 32 mm and 1× GL 25 male, total length 110 mm
- Pk:1
Dane techniczne
Informacje o tym produkcie
T-shaped connector made of static dissipative PE-EX.
Lateral connections with union nut and clamp ring to insert and clamp b.safe waste pipes with Ø 32 mm. Central connection with male thread GL 25 to mount e.g. b.safe waste tubes.