Order Entry
Poland
ContactUsLinkComponent
Connector for waste pipes, b.safe
Connector for waste pipes, b.safe
  BOHLR1172-01
 :  Bohlender
undefined
Connector for waste pipes, b.safe
  BOHLR1172-01
 :  Bohlender
  • Description:
    Connector for waste pipes, PE, 2× clamp connection Ø 32 mm
  • Pk:
    1
Straight connector made of static dissipative PE-EX.
With lock nuts and clamp rings on both sides for fixing and clamping b.safe waste pipes with Ext.Ø 32 mm.