Order Entry
Poland
Orders LinkContactUsLinkComponent
Hydrogen generators, Precision SL 100
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
Hydrogen generators, Precision SL 100
   Sustainable Options Available
Supplier:   PEAK SCIENTIFIC
Hydrogen generators, Precision SL 100
Supplier:  PEAK SCIENTIFIC
   Sustainable Options Available
Certificates
Hydrogen generators, Precision SL 100
Hydrogen generators, Precision SL 100
12 Options Available Below
Product Details & Documents
The smallest hydrogen generator for GC-FID, Precision hydrogen SL has been developed to offer a streamlined laboratory-grade hydrogen gas solution for GC flame detectors at a purity of 99,9995%. Precision hydrogen SL is safe, simple to use, easy to maintain and takes up minimal bench space.
  • 2 models – 100 and 200 ml
  • 99,9995% purity at 6,89 bar
  • One-button start-up and shut-down
  • Easy end-user managed servicing
  • Optional auto-water fill or pressurised water fill
  • Auto-shut-down fail-safe feature
  • Choice of colours available: Black or white
  • 2 year comprehensive warranty
With an impressive, streamlined form factor, Precision Hydrogen SL produces hydrogen at up to 100 ml/min with no compromise on safety. A range of features ensure that Precision SL will produce hydrogen safely, storing a minimal volume of gas compared to cylinders. This generator also comes in a choice of black or white and includes a 2 year warranty as standard.
Certifications: CE marked, FCC and CSA compliant
Product Family Options
Product Information
  • Availability
    Price
  • PEAK3302583
    De-ioniser cartridge hydrogen
    De-ioniser cartridge hydrogen
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 821 zł
    821 zł
    1 * 1 SZT
       821 zł
    Specifications:
    More
    Specifications
    Cat. No.
    PEAK3302583
  • 564-0286
    Precision SL 100 hydrogen generator, manual water fill (white)
    Precision SL 100 hydrogen generator, manual water fill (white)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 29 280 zł
    29 280 zł
    1 * 1 SZT
       29 280 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0286
  • 564-0287
    Precision SL 100 hydrogen generator, auto-water fill, (white)
    Precision SL 100 hydrogen generator, auto-water fill, (white)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 30 990 zł
    30 990 zł
    1 * 1 SZT
       30 990 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0287
  • 564-0288
    Precision SL 100 hydrogen generator, pressurised water fill (white)
    Precision SL 100 hydrogen generator, pressurised water fill (white)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 32 700 zł
    32 700 zł
    1 * 1 SZT
       32 700 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0288
  • 564-0289
    Precision SL 100 hydrogen generator, manual water fill (black)
    Precision SL 100 hydrogen generator, manual water fill (black)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 29 280 zł
    29 280 zł
    1 * 1 SZT
       29 280 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0289
  • 564-0290
    Precision SL 100 hydrogen generator, auto-water fill (black)
    Precision SL 100 hydrogen generator, auto-water fill (black)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 30 990 zł
    30 990 zł
    1 * 1 SZT
       30 990 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0290
  • 564-0291
    Precision SL 100 hydrogen generator, pressurised water fill (black)
    Precision SL 100 hydrogen generator, pressurised water fill (black)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 32 700 zł
    32 700 zł
    1 * 1 SZT
       32 700 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0291
  • 564-0292
    Precision SL 200 hydrogen generator, manual water fill (white)
    Precision SL 200 hydrogen generator, manual water fill (white)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 33 050 zł
    33 050 zł
    1 * 1 SZT
       33 050 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0292
  • 564-0293
    Precision SL 200 hydrogen generator, auto-water fill (white)
    Precision SL 200 hydrogen generator, auto-water fill (white)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 34 370 zł
    34 370 zł
    1 * 1 SZT
       34 370 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0293
  • 564-0294
    Precision SL 200 hydrogen generator, pressurised water fill (white)
    Precision SL 200 hydrogen generator, pressurised water fill (white)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 36 080 zł
    36 080 zł
    1 * 1 SZT
       36 080 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0294
  • 564-0296
    Precision SL 200 hydrogen generator, auto-water fill (black)
    Precision SL 200 hydrogen generator, auto-water fill (black)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 34 370 zł
    34 370 zł
    1 * 1 SZT
       34 370 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0296
  • 564-0297
    Precision SL 200 hydrogen generator, pressurised water fill (black)
    Precision SL 200 hydrogen generator, pressurised water fill (black)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 36 080 zł
    36 080 zł
    1 * 1 SZT
       36 080 zł
    Specifications:
    More
    Specifications
    Cat. No.
    564-0297
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Desiccant capsule
    Desiccant capsule
    PEAK3302581
    Log in for availability
    1 * 1 SZT - 520 zł