Vecuronium bromide 98%
ACRO461972500
:
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
- Appearance:White to almost white crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤ 4% (105°C)
- Titration with HClO4:97.5 to 102.5% (dry substance)
- Specific optical rotation:+30.5° to +35° (20°C, 589nm) (1% in 0.05M HCl)
:
:637.74 g/mol
:13.10003
:235°C
:256-723-9
:GHS07
:Ambient
:50700-72-6
:C₃₄H₅₇BrN₂O₄
:MFCD00867762