Order Entry
Poland
ContactUsLinkComponent
Ammonia solution 20 - 22% HiPerSolv CHROMANORM® for LC-MS
Irritant
Corrosive
Environment
Ammonia solution 20 - 22% HiPerSolv CHROMANORM® for LC-MS
Catalog # 88439.154
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Environment
Ammonia solution 20 - 22% HiPerSolv CHROMANORM® for LC-MS
Catalog # 88439.154
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Assay (GC):
    20 - 22%
  • Evaporation residue:
    max. 0,0002%
  • Ca (Calcium):
    max. 2 ppb
  • K (Potassium):
    max. 2 ppb
  • Mg (Magnesium):
    max. 2 ppb
  • Na (Sodium):
    max. 10 ppb
  • Suitable for LC-MS:
    Passes test

Specification Test Results

About this item

 :
 :35.05 g/mol
ADR:8,III
 :Packaged under nitrogen.
UN:2672
 :GHS07
 :GHS05
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650