VWR®, Pożywki do hodowli komórkowych, Schneider's Drosophila medium
Catalog # 392-0419
Supplier: VWR Chemicals
CAS Number:
Specifications
- Format mediów:Płyn
- Ilość:500 mL
- Opis:Schneider's Drosophila medium
- Wapń:Yes
- Magnez:Yes
- HEPES:No
- Wodorowęglan sodu:Yes
- Pirogronian sodu:No
- Czerwień fenolowa:No
- Stabilna glutamina:No
- L-Glutamina:Yes
- Media Base:Schneider's Insect Medium
Specifications
About this item
Schneider's Drosophila medium promotes rapid growth of both primary and existing cell cultures derived from Drosophila spp and several other dipterans when supplemented with 5 to 20% heat-inactivated foetal bovine serum.