Order Entry
Poland
ContactUsLinkComponent
Affinity removal spin cartridges HSA
Affinity removal spin cartridges HSA
Catalog # HEWL5188-5334
Supplier:  Agilent
CAS Number:  
Affinity removal spin cartridges HSA
Catalog # HEWL5188-5334
Supplier:  Agilent
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Affinity removal system spin cartridge; human albumin, 0,45 ml
  • Seq:
    01
Specifications
About this item
The affinity removal spin cartridge – human albumin is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Compatible with standard benchtop microcentrifuges and Luer-lok syringes
  • Serum load volume is 65 µl per run
  • Optimise sample recovery by utilising our rapid, single injection protocols and optimised buffers
Removal of albumin improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.