Order Entry
Poland
ContactUsLinkComponent
Pump kits
Pump kits
Catalog # HEWL5067-5378
Supplier:  Agilent
undefined
Pump kits
Catalog # HEWL5067-5378
Supplier:  Agilent
Specifications
  • Description:
    Inlet tubing kit, for 1260 infinity quaternary pump VL
  • Pk:
    1
Specifications
About this item
A variety of high quality pump kits for LC systems and applications, containing all fittings and connections required.
  • To generally clean the vacuum degasser tubing, flush the system with isopropanol
  • Flush the degasser with water after using buffers
  • Speed solvent changes by drawing solvent through the degasser and tubing with syringe adapter kit
Pump start-up kits for RRLC pumps and nanoflow LC systems are provided, along with seal wash kits for active, continuous or preventative maintenance seal washing purposes. Pump kits include all relevant fittings, tubing, valves, seals and other components required to produce the highest levels of performance in all LC pumps and systems.