Order Entry
Poland
ContactUsLinkComponent
 
Copper(II) tartrate hydrate ≥99.9% (trace metals basis), Sigma-Aldrich®
Catalog # SIAL514071-50G
Supplier:  Merck
CAS Number:  
Copper(II) tartrate hydrate ≥99.9% (trace metals basis), Sigma-Aldrich®
Catalog # SIAL514071-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :211.62 g/mol (anhy)
Melting Pt:~275 °C (dec., lit.)
 :M-Clarity™ quality level = MQ200
CAS Number:946843-80-7
 :815-82-7
Formula:C₄H₄CuO₆·xH₂O
MDL Number:MFCD00150650