Order Entry
Poland
ContactUsLinkComponent
Single element standard for ICP-MS, aluminum
Single element standard for ICP-MS, aluminum
Catalog # HEWL5190-8561
Supplier:  Agilent
CAS Number:  
undefined
Single element standard for ICP-MS, aluminum
Catalog # HEWL5190-8561
Supplier:  Agilent
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pk:
    1 mL
  • Pk Info:
    100 ml
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item

It offers a comprehensive range of single and multi-element standards, tuning solutions, ionization buffers and environmental standards for Atomic Spectroscopy.

  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations

All standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.

Delivery: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Packaging: All standards packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.