J&W HP-PLOT Al₂O₃ KCl GC columns
Supplier: Agilent
Certificates
About this item
Agilent J&W HP-PLOT Al₂O₃ KCl is the least polar alumina phase and is deactivated with sodium chloride.
- Least polar alumina phase
- Low retention of olefins relative to comparable paraffin
- Recommended phase for many ASTM methods
It is the standard column choice for light hydrocarbon analysis, particularly C1 to C8 hydrocarbon isomers. Excellent for the quantification of dienes, especially propadiene and butadiene from ethylene and propylene streams.
Frequently Bought Together