Order Entry
Poland
ContactUsLinkComponent
 
PD-123319 ditrifluoroacetate, hydrate ≥98% (by HPLC), Sigma-Aldrich®
Catalog # SIALP186-10MG
Supplier:  Merck
CAS Number:  
PD-123319 ditrifluoroacetate, hydrate ≥98% (by HPLC), Sigma-Aldrich®
Catalog # SIALP186-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :736.66 g/mol (anhy)
CAS Number:136676-91-0
 :130663-39-7
Formula:C₃₁H₃₂N₄O₃·2C₂HF₃O₂·xH₂O
MDL Number:MFCD11046022