Human recombinant Deoxyribonuclease I
Catalog # APOSBITP1317-500IU
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1
- Enzyme type:Recombinant
- Species:Human
- Storage conditions:Ambient
- MDL No.:MFCD00130918
- Enzyme name:Deoxyribonuclease I
- Enzyme synonyms:DNase
- Pk Info:500 U