Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-CCL3 Mouse Antibody
Anti-CCL3 Mouse Antibody
Anti-CCL3 Mouse Antibody
Catalog # APOSBITP1994-1MG
Supplier:
Apollo Scientific
Expand image
Anti-CCL3 Mouse Antibody
Catalog # APOSBITP1994-1MG
Supplier:
Apollo Scientific
Specifications
Antigen name:
Macrophage Inflammatory Protein-1a
Antigen symbol:
CCL3
Host:
Mouse
Reactivity:
Human
Pk:
1 mg
Specifications
About this item
Product Details
Product Details