Order Entry
Poland
ContactUsLinkComponent
Anti-FAS A Mouse Antibody
Anti-FAS A Mouse Antibody
  APOSBITP1968-1MG
Anti-FAS A Mouse Antibody
  APOSBITP1968-1MG

 

  • Antigen name:
    FAS (CD95) Activating antibody
  • Antigen symbol:
    FAS A
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg