Order Entry
Poland
ContactUsLinkComponent
Bovine Recombinant Leptin
Bovine Recombinant Leptin
Catalog # APOSBITP1199-1MG
Supplier:  Apollo Scientific
CAS Number:  
Bovine Recombinant Leptin
Catalog # APOSBITP1199-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Bovine
  • Protein/peptide name:
    Leptin

Specifications

About this item